ADVH303BA2E7 AChemBlock

2-methyl-6-(trifluoromethyl)-1H-indole

  • MDL number MFCD16875985
  • Cas Number : 57330-48-0
  • Molecular Weight : 199.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-6-trifluoromethyl-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55370
Availability details for
Quantity
  • Availability
  • Add to Cart