ADVH303BA2F0 AChemBlock

methyl 5-fluoro-1H-indole-6-carboxylate

  • MDL number MFCD16875726
  • Cas Number : 1227268-61-2
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-fluoro-1h-indole-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55379
Availability details for
Quantity
  • Availability
  • Add to Cart