ADVH303BA305 AChemBlock

6-fluoro-4-methoxy-1H-indole

  • MDL number MFCD07781565
  • Cas Number : 885521-02-8
  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-4-methoxy-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55400
Availability details for
Quantity
  • Availability
  • Add to Cart