ADVH303BA3A1 AChemBlock

4,6-dichloro-1,3-dihydro-2H-pyrrolo[3,2-c]pyridin-2-one

  • MDL number MFCD18250905
  • Cas Number : 1260672-68-1
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-1-3-dihydro-2h-pyrrolo-3-2-c-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55575
Availability details for
Quantity
  • Availability
  • Add to Cart