ADVH303BA3A3 AChemBlock

4-chloro-6-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD10697160
  • Cas Number : 30129-53-4
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-methyl-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55577
Availability details for
Quantity
  • Availability
  • Add to Cart