ADVH303BA431 AChemBlock

methyl 3,3-dimethyl-2-oxoindoline-6-carboxylate

  • MDL number MFCD28502331
  • Cas Number : 1638765-21-5
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-3-dimethyl-2-oxoindoline-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55730
Availability details for
Quantity
  • Availability
  • Add to Cart