ADVH303BA441 AChemBlock

5-bromoimidazo[1,2-a]pyridin-2-amine

  • MDL number MFCD20502277
  • Cas Number : 1350738-82-7
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromoimidazo-1-2-a-pyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55746
Availability details for
Quantity
  • Availability
  • Add to Cart