ADVH303BA442 AChemBlock

tert-butyl (S)-(5-oxopyrrolidin-3-yl)carbamate

  • MDL number MFCD20257110
  • Cas Number : 672883-23-7
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-s-5-oxopyrrolidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55747
Availability details for
Quantity
  • Availability
  • Add to Cart