ADVH303BA538 AChemBlock

tert-butyl 8-oxo-2-oxa-5-azaspiro[3.5]nonane-5-carboxylate

  • MDL number MFCD31743156
  • Cas Number : 1934939-07-7
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-8-oxo-2-oxa-5-azaspiro-3-5-nonane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56009
Availability details for
Quantity
  • Availability
  • Add to Cart