ADVH303BA545 AChemBlock

spiro[3.3]heptane-2,6-dione

  • MDL number MFCD24689331
  • Cas Number : 20061-23-8
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/spiro-3-3-heptane-2-6-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56023
Availability details for
Quantity
  • Availability
  • Add to Cart