ADVH303BA5A2 AChemBlock

5-oxa-2-azaspiro[3.4]octan-7-ol

  • MDL number MFCD30802728
  • Cas Number : 1613363-18-0
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-oxa-2-azaspiro-3-4-octan-7-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56124
Availability details for
Quantity
  • Availability
  • Add to Cart