ADVH303BA5B9 AChemBlock

(3-(fluoromethyl)oxetan-3-yl)methanol

  • MDL number MFCD29968532
  • Cas Number : 61729-10-0
  • Molecular Weight : 120.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoromethyl-oxetan-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56148
Availability details for
Quantity
  • Availability
  • Add to Cart