ADVH303BA5E0 AChemBlock

tert-butyl (7-oxospiro[3.5]nonan-2-yl)carbamate

  • MDL number MFCD30749720
  • Cas Number : 2168870-40-2
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-oxospiro-3-5-nonan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56189
Availability details for
Quantity
  • Availability
  • Add to Cart