ADVH303BA6C6 AChemBlock

7-bromo-1H-indazole-3-carboxylic acid

  • MDL number MFCD07371559
  • Cas Number : 885278-71-7
  • Molecular Weight : 241.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1h-indazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56418
Availability details for
Quantity
  • Availability
  • Add to Cart