ADVH303BA6DE AChemBlock

5H-pyrrolo[3,2-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD17013012
  • Cas Number : 857204-03-6
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5h-pyrrolo-3-2-d-pyrimidine-6-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56450
Availability details for
Quantity
  • Availability
  • Add to Cart