ADVH303BA6F9 AChemBlock

2-(tert-butoxycarbonyl)hexahydrocyclopenta[c]pyrrole-3a(1H)-carboxylic acid

  • MDL number MFCD20230745
  • Cas Number : 1314394-96-1
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butoxycarbonyl-hexahydrocyclopenta-c-pyrrole-3a-1h-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56480
Availability details for
Quantity
  • Availability
  • Add to Cart