ADVH303BA720 AChemBlock

tert-butyl 3-(iodomethyl)-3-methylazetidine-1-carboxylate

  • MDL number MFCD23106025
  • Cas Number : 1408074-76-9
  • Molecular Weight : 311.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-iodomethyl-3-methylazetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56524
Availability details for
Quantity
  • Availability
  • Add to Cart