ADVH303BA75B AChemBlock

7-(tert-butoxycarbonyl)-1-oxa-2,7-diazaspiro[4.4]non-2-ene-3-carboxylic acid

  • MDL number MFCD12198541
  • Cas Number : 1160247-02-8
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-tert-butoxycarbonyl-1-oxa-2-7-diazaspiro-4-4-non-2-ene-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56589
Availability details for
Quantity
  • Availability
  • Add to Cart