ADVH303BA761 AChemBlock

tert-butyl 1-oxa-4,8-diazaspiro[5.5]undecane-4-carboxylate

  • MDL number MFCD11227069
  • Cas Number : 1160247-06-2
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-oxa-4-8-diazaspiro-5-5-undecane-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56595
Availability details for
Quantity
  • Availability
  • Add to Cart