ADVH303BA7AC AChemBlock

(2,6-dichloropyrimidin-4-yl)methanol

  • MDL number MFCD28108065
  • Cas Number : 321329-01-5
  • Molecular Weight : 179.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloropyrimidin-4-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56672
Availability details for
Quantity
  • Availability
  • Add to Cart