ADVH303BA7C3 AChemBlock

4-chloro-3-methoxypyridin-2-amine

  • MDL number MFCD18415420
  • Cas Number : 1261452-97-4
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-3-methoxypyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56241
Availability details for
Quantity
  • Availability
  • Add to Cart