ADVH303BA8A1 AChemBlock

rel-tert-butyl (7R,8aS)-7-hydroxyhexahydropyrrolo[1,2-a]pyrazine-2(1H)-carboxylate

  • Cas Number : 1204603-42-8
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/rel-tert-butyl-7r-8as-7-hydroxyhexahydropyrrolo-1-2-a-pyrazine-2-1h-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56941
Availability details for
Quantity
  • Availability
  • Add to Cart