ADVH303BA8E7 AChemBlock

2-((1R,3S)-3-hydroxycyclopentyl)acetic acid

  • Cas Number : 876605-24-2
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1r-3s-3-hydroxycyclopentyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q57015
Availability details for
Quantity
  • Availability
  • Add to Cart