ADVH303BA93A AChemBlock

(7-oxaspiro[3.5]nonan-2-yl)methanamine hydrochloride

  • MDL number MFCD31743244
  • Cas Number : 2250243-27-5
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-oxaspiro-3-5-nonan-2-yl-methanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q57103
Availability details for
Quantity
  • Availability
  • Add to Cart