ADVH303BA94C AChemBlock

tert-butyl (3-hydroxy-1-methylcyclopentyl)carbamate

  • MDL number MFCD28612286
  • Cas Number : 1780760-71-5
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-hydroxy-1-methylcyclopentyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q57124
Availability details for
Quantity
  • Availability
  • Add to Cart