ADVH303BA980 AChemBlock

(S)-6-oxaspiro[2.5]octane-1-carboxylic acid

  • Cas Number : 909406-97-9
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/s-6-oxaspiro-2-5-octane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q57179
Availability details for
Quantity
  • Availability
  • Add to Cart