ADVH303BA985 AChemBlock

methyl 4-oxobicyclo[4.1.0]heptane-1-carboxylate

  • MDL number MFCD30802966
  • Cas Number : 1535963-64-4
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-oxobicyclo-4-1-0-heptane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q57184
Availability details for
Quantity
  • Availability
  • Add to Cart