ADVH303BB524 AChemBlock

1-(imidazo[1,2-a]pyridin-6-yl)ethan-1-one

  • MDL number MFCD10696290
  • Cas Number : 944905-12-8
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-imidazo-1-2-a-pyridin-6-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60079
Availability details for
Quantity
  • Availability
  • Add to Cart