ADVH303BB5B2 AChemBlock

6-methyl-2-oxoindoline-3-carbaldehyde

  • MDL number MFCD09971851
  • Cas Number : 845655-53-0
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-2-oxoindoline-3-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60222
Availability details for
Quantity
  • Availability
  • Add to Cart