ADVH303BB5CA AChemBlock

tetrahydro-2H-pyran-2-carbaldehyde

  • MDL number MFCD08235010
  • Cas Number : 19611-45-1
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tetrahydro-2h-pyran-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60246
Availability details for
Quantity
  • Availability
  • Add to Cart