ADVH303BB68B AChemBlock

(3R,4R)-3-methyl-2-oxa-8-azaspiro[4.5]decan-4-amine dihydrochloride

  • Cas Number : 2227197-87-5
  • Molecular Weight : 243.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-4r-3-methyl-2-oxa-8-azaspiro-4-5-decan-4-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61354
Availability details for
Quantity
  • Availability
  • Add to Cart