ADVH303BB744 AChemBlock

3-chloro-4-methoxypicolinic acid

  • MDL number MFCD18257362
  • Cas Number : 1256822-07-7
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-4-methoxypicolinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60578
Availability details for
Quantity
  • Availability
  • Add to Cart