ADVH303BB825 AChemBlock

2-isopropoxypyrimidine-5-carboxylic acid

  • MDL number MFCD16657353
  • Cas Number : 1072855-45-8
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-isopropoxypyrimidine-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60807
Availability details for
Quantity
  • Availability
  • Add to Cart