ADVH303BB86F AChemBlock

5-methoxy-2-methylnicotinic acid

  • MDL number MFCD18254599
  • Cas Number : 1174402-76-6
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-2-methylnicotinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q60882
Availability details for
Quantity
  • Availability
  • Add to Cart