ADVH303BB91C AChemBlock

tert-butyl 3-amino-5-fluoropiperidine-1-carboxylate

  • MDL number MFCD21601921
  • Cas Number : 1356342-41-0
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-5-fluoropiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61063
Availability details for
Quantity
  • Availability
  • Add to Cart