ADVH303BBA45 AChemBlock

tert-butyl 4,4-difluoro-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD31431542
  • Cas Number : 2137836-39-4
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-4-difluoro-1-oxa-6-azaspiro-2-5-octane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61367
Availability details for
Quantity
  • Availability
  • Add to Cart