ADVH303BBB7F AChemBlock

ethyl 3-chloro-1H-pyrazole-4-carboxylate

  • MDL number MFCD28050408
  • Cas Number : 1393667-83-8
  • Molecular Weight : 174.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-chloro-1h-pyrazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61544
Availability details for
Quantity
  • Availability
  • Add to Cart