ADVH303BBBF1 AChemBlock

tert-butyl ((3R,5R)-5-methylpyrrolidin-3-yl)carbamate

  • MDL number MFCD19687845
  • Cas Number : 1932651-04-1
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-5r-5-methylpyrrolidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61666
Availability details for
Quantity
  • Availability
  • Add to Cart