ADVH303BBC33 AChemBlock

1-(tert-butyl) 3-methyl (S)-3-methoxypyrrolidine-1,3-dicarboxylate

  • MDL number MFCD28502671
  • Cas Number : 1184174-08-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-3-methyl-s-3-methoxypyrrolidine-1-3-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61736
Availability details for
Quantity
  • Availability
  • Add to Cart