ADVH303BBC4D AChemBlock

methyl (1S,2S)-2-((tert-butoxycarbonyl)amino)cyclopent-3-ene-1-carboxylate

  • MDL number MFCD29035276
  • Cas Number : 1272666-21-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1s-2s-2-tert-butoxycarbonyl-amino-cyclopent-3-ene-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q61768
Availability details for
Quantity
  • Availability
  • Add to Cart