ADVH303BBDAE AChemBlock

2-(3-amino-1-methyl-1H-pyrazol-5-yl)propan-2-ol

  • MDL number MFCD31743604
  • Cas Number : 1649468-56-3
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-amino-1-methyl-1h-pyrazol-5-yl-propan-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62137
Availability details for
Quantity
  • Availability
  • Add to Cart