ADVH303BBDF0 AChemBlock

7-fluoro-1H-indole-5-carbonitrile

  • MDL number MFCD04037880
  • Cas Number : 883500-88-7
  • Molecular Weight : 160.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-fluoro-1h-indole-5-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62205
Availability details for
Quantity
  • Availability
  • Add to Cart