ADVH303BBE48 AChemBlock

6-(trifluoromethoxy)nicotinic acid

  • MDL number MFCD17677182
  • Cas Number : 940895-85-2
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethoxy-nicotinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62295
Availability details for
Quantity
  • Availability
  • Add to Cart