ADVH303BBFDE AChemBlock

spiro[2.4]heptan-5-one

  • MDL number MFCD24690087
  • Cas Number : 19740-31-9
  • Molecular Weight : 110.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/spiro-2-4-heptan-5-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62619
Availability details for
Quantity
  • Availability
  • Add to Cart