ADVH303BBFE5 AChemBlock

imidazo[1,2-a]pyridine-6-sulfonyl chloride

  • MDL number MFCD28606679
  • Cas Number : 1444886-02-5
  • Molecular Weight : 216.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-2-a-pyridine-6-sulfonyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62436
Availability details for
Quantity
  • Availability
  • Add to Cart