ADVH303BC06C AChemBlock

(R)-3-fluoropyrrolidine-1-sulfonyl chloride

  • MDL number MFCD30533071
  • Cas Number : 1411774-27-0
  • Molecular Weight : 187.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-3-fluoropyrrolidine-1-sulfonyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62769
Availability details for
Quantity
  • Availability
  • Add to Cart