ADVH303BC074 AChemBlock

2-ethynyloxetane

  • MDL number MFCD30803399
  • Cas Number : 33869-43-1
  • Molecular Weight : 82.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-ethynyloxetane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62777
Availability details for
Quantity
  • Availability
  • Add to Cart