ADVH303BC0C2 AChemBlock

(3S,6R)-1-(tert-butoxycarbonyl)-6-methylpiperidine-3-carboxylic acid

  • MDL number MFCD30163414
  • Cas Number : 1253200-92-8
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3s-6r-1-tert-butoxycarbonyl-6-methylpiperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62858
Availability details for
Quantity
  • Availability
  • Add to Cart