ADVH303BC0D2 AChemBlock

(3R,5S)-5-(((benzyloxy)carbonyl)amino)-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid

  • MDL number MFCD30803535
  • Cas Number : 1145747-62-1
  • Molecular Weight : 378.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-5s-5-benzyloxy-carbonyl-amino-1-tert-butoxycarbonyl-piperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62875
Availability details for
Quantity
  • Availability
  • Add to Cart