ADVH303BC0E0 AChemBlock

4,4-difluoro-1-methylpiperidin-3-ol

  • MDL number MFCD30803558
  • Cas Number : 1785484-23-2
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-1-methylpiperidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62890
Availability details for
Quantity
  • Availability
  • Add to Cart